Imprimer
Trousers, Fristads® PR54-220, Design B, black
Fournisseur: Fristads Kansas Group
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Fristads®
113-01540EA 0 EUR
113-01540 113-03048 113-03047 113-01541 113-03046 113-03045 113-03044 113-03043 113-03042 113-03041 113-01427 113-01428 113-01425 113-01426 113-01542 113-03049 113-01543 113-01539 113-03040 113-01430 113-03059 113-01431 113-03058 113-03057 113-03056 113-03055 113-03054 113-03053 113-03052 113-01438 113-01439 113-01436 113-01437 113-01434 113-01435 113-01432 113-01433 113-01429 113-03051 113-03050 113-01441 113-03069 113-01442 113-03068 113-01440 113-03067 113-03066 113-03065 113-03064 113-03063 113-01449 113-01447 113-01448 113-01445 113-01446 113-01443 113-01444 113-03062 113-03061 113-03060 113-01452 113-01453 113-03079 113-01450 113-03078 113-01451 113-03077 113-03076 113-03075 113-03074 113-01458 113-01459 113-01456 113-01457 113-01454 113-01455 113-03073 113-03072 113-03071 113-03070 113-01463 113-01464 113-01461 113-01462 113-01460 113-01504 113-01505 113-01469 113-01502 113-01503 113-01500 113-01467 113-01501 113-01468 113-01465 113-01466 113-03082 113-03081 113-03080 113-03015 113-01474 113-01475 113-03014 113-03013 113-01472 113-03012 113-01473 113-03011 113-01470 113-01471 113-01515 113-01516 113-01513 113-01514 113-03019 113-01511 113-01478 113-01512 113-03018 113-01479 113-03017 113-01476 113-01510 113-03016 113-01477 113-01508 113-01509 113-01506 113-01507 113-03026 113-01485 113-03025 113-01486 113-01483 113-03024 113-03023 113-01484 113-03022 113-01481 113-01482 113-03021 113-03020 113-01480 113-01526 113-01527 113-01524 113-01525 113-01489 113-01522 113-03029 113-01523 113-01520 113-01487 113-03028 113-01488 113-03027 113-01521 113-01519 113-01517 113-01518 113-01496 113-03037 113-03036 113-01497 113-01530 113-01494 113-03035 113-01495 113-03034 113-03033 113-01492 113-01493 113-03032 113-01490 113-03031 113-03030 113-01491 113-01537 113-01538 113-01535 113-01536 113-01533 113-01534 113-01531 113-03039 113-01498 113-01499 113-01532 113-03038 113-01528 113-01529
Trousers, Fristads® PR54-220, Design B, black
Tenue de travail
Thanks to their durability and comfort, these trousers are perfectly suitable for use in garages as well as in industrial and service companies. The material is made of 65% polyester and 35% cotton. The inside is brushed and provides a comfortable cotton feel. The outside is very hardwearing and does not bleach, even after countless washes.
  • Two front and back pockets
  • Two leg pockets - one with flap, telephone pocket, pen pocket, tool bags
  • Adjustable waist, plastic snap buttons
  • Colour accents on pocket edges and leg pocket flap
Weight: 260 g/m²
Basic colour: black