Imprimer
Sac fond rond renforcé
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
129-0491EA 197 EUR
PBIB89211
Sac fond rond renforcé
Blender bags made of high strength plastic material, transparent, for use in circulator homogenisers 'stomacher 400 circulator, 'stomacher 400' or similar.
  • Tear-off top seal to guarantee sterility of every bag
  • Round bottom to avoid stagnancy in the corners
  • Reinforced bottom with double-seal to prevent leakage
  • Graduated to 400 ml
  • Label for identification of the sample according to GLP and lot number printed on every bag