Imprimer
Grid and channel trays, Refrig/Arrangers®
Fournisseur: Spectrum
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SPEC183550EA 143 EUR
SPEC183550 SPEC183556 SPEC183554
Grid and channel trays, Refrig/Arrangers®
Made of polystyrene.
  • Lightweight and sturdy trays
  • Ideal for freezer storage as low as −90 °C
  • Secondary containment for spills
Suitable for the safe and convenient transportation and storage of glass vials and small bottles.