Imprimer
Screws with countersunk, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOHLH1124-22EA 23.8 EUR
BOHLH1124-22 BOHLH1124-14 BOHLH1124-26 BOHLH1124-18
Screws with countersunk, BOLA
Fixations Accouplements
PTFE, white.
  • Temperature resistance: from -200 °C to +250 °C
  • Universal chemical resistance
  • FDA conform
  • Similar to DIN 963/DIN EN ISO 2009