Imprimer
Aeration cannula
Fournisseur: B. Braun
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BRAU4190050EA 305 EUR
BRAU4190050
Aeration cannula
Canules
Aeration cannula maintains a bacteria-retentive venting of noncollapsing plastic containers.
Suitable for applications in anesthesiae, intensive care, preparation of cytotoxics and general attention.
Cdt: 100 units per box in individual blisters.