Imprimer
Titrator, automatic sample changer, TW alpha plus
Fournisseur: SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SCOI1007305EA 11850 EUR
SCOI1007305
Titrator, automatic sample changer, TW alpha plus
This sample changer with four sample racks for up to 24 samples and titration head fittings for a variety beaker and titrator vessels enables users to titrate in series with automatic sample changing.
  • Changer can be controlled via TitroLine alpha plus titrator or via a PC with the TitriSoft software
  • With integrated magnetic stirrer to stir the samples from 'below' – alternatively, a rod stirrer can be used
  • Higher flexibility due to exchangeable sample racks
  • Various sample plates from 12 to 24 positions for standard bechers acc. to DIN
Informations de livraison: Supplied with integrated magnetic stirrer and connection cable TZ 1581 for rod stirrer.