Imprimer
Votre recherche pour: Tests sur les endoxines
13  résultat(s)
SearchResultCount:"13"
Sort Results
Vue liste Nouvelle Vue
Evaluer les résultats de votre recherche
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: The Toxinometer® ET-7000 is a computer-operated kinetic incubating tube reader, designed to be exceptionally user-friendly. Our state-of-the-art expan...
New Product
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: The PYROSTAR™ ES-F series of reagents represent our uniquely endotoxin specific line of Limulus Amebocyte Lysate products, which are formulated to be ...
New Product
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: The PYROSTAR™ ES-F series of reagents represent our uniquely endotoxin specific line of Limulus Amebocyte Lysate products, which are formulated to be ...
New Product
Référence Produit: (WAKF294-36731)
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: PYROSTAR Neo is a new endotoxin detection reagent developed by recombinant technology.
UOM: 1 * 2,7 mL
New Product
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: The Limulus PS Single Test is composed of LAL endotoxin-specific single-test vials and an affinity resin suspension called PyroSep™, designed to overc...
New Product
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: The Limulus Color KY Series are quantitative kinetic chromogenic assays (KCA) that are endotoxin-specific and unreactive to (1→3)-β-D-glucan. This ser...
New Product
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: The LPS for Endotoxin Indicator is composed of E. coli O55:B5 (>100,000 EU/vial) and serves as a stock solution to prepare multiple endotox...
New Product
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: The Limulus Color KY Series are quantitative kinetic chromogenic assays (KCA) that are endotoxin-specific and unreactive to (1→3)-β-D-glucan. This ser...
New Product
Fournisseur: FUJIFILM WAKO CHEMICALS
Description: The Control Standard Endotoxin (CSE) is a lyophilised product composed of a 500 ng vial of endotoxin, purified from E. coli UKT-B strain that s...
New Product
Fournisseur: Thermo Fisher Scientific
Description: The Thermo Scientific Pierce™ Chromogenic endotoxin Quant kit is a highly sensitive endpoint assay that accurately measures and detects endotoxin (lip...
Fournisseur: Genscript
Description: ToxinSensor™ single tests kit with standard is designed to be a qualitative In Vitro end-point endotoxin test for human and animal parenteral drugs, b...
Référence Produit: (GENSL00351)
Fournisseur: Genscript
Description: ToxinSensor™ Gel Clot Endotoxin Assay Kit is intended as an in vitro end-product endotoxin test for human and animal parenteral drugs, biologic...
UOM: 1 * 1 KIT
Fournisseur: Genscript
Description: ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit is designed to be a quantitative In vitro end-point endotoxin test for human and animal parent...
Prix sur demande
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Le stock de cet article est limité mais peut être disponible dans un entrepôt proche de vous. Merci de vous assurer que vous êtes connecté sur le site afin que le stock disponible soit affiché. Si l'call est toujours affiché et vous avez besoin d'aide, s'il vous plaît appelez-nous au 01 45 14 89 12.
Ces articles ne peuvent pas être ajoutés à votre panier en raison des exigences réglementaires. Veuillez envoyer un e-mail à webshop.fr@vwr.com
Veuillez noter qu'une autorisation peut être nécessaire pour commander ce produit. Un représentant de VWR vous contactera si votre commande le nécessite.
Ce produit a été bloqué par votre entreprise. Veuillez s'il vous plait contacter votre service achats pour plus d'informations.
Le produit original n'est plus disponible. Le remplacement représenté est disponible
Ce produit n'est plus disponible. Des alternatives peuvent être disponibles en faisant une recherche avec la référence VWR ci-dessus. Si vous avez besoin d'aide, s'il vous plaît appelez le Service Clients au 0825 02 30 30 (0,18 € TTC/mn).
1 - 13 of 13
no targeter for Bottom