Joint clips for conical joints, PTFE/steel spring, Quickfit®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Quickfit®
BIBBGC12EA 50.9 GBP
BIBBGC12 BIBBGC45 201-8560 201-3027 201-8559 201-8558 201-3026 201-8557 201-3025
Joint clips for conical joints, PTFE/steel spring, Quickfit®
Clamps NS Joint Clamps
Clips for retaining glassware having Quickfit® conical joints. Manufactured from PTFE with encapsulated steel spring to provide a firm grip.
  • Excellent chemical resistance
  • Can be used at temperatures up to 250 °C
  • Clips are numbered with joint size for easy identification