Imprimer
Laboratory/media bottles, narrow neck, borosilicate glass, plastic-coated, without screw cap and pouring ring, BIBBY®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BIBB1518/16EA 466 EUR
BIBB1518/16 BIBB1518/18 BIBB1518/08 BIBB1518/10 BIBB1518/06 BIBB1518/04 BIBB1518/12
Laboratory/media bottles, narrow neck, borosilicate glass, plastic-coated, without screw cap and pouring ring, BIBBY®
Flacons Flacons en verre
PYREX® borosilicate glass without cap or pouring ring.
  • Clear plastic coated to protect against mechanical breakage
  • Reduces the hazard of flying glass and liquid splashes in the event of breakage
  • Coating withstands steam sterilisation up to 135 °C
Printed with retrace code; downloadable batch certificate.